Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001197295.1 | NP_001184224.1 | 6 | 1393 | ||
NM_001197296.1 | NP_001184225.1 | 6 | 1393 | |||
NM_001393.3 | NP_001384.1 | 6 | 1459 | |||
XM_017014376.1 | XP_016869865.1 | 5 | 1299 | |||
GenBank mRNA | AB011792.1 | - | 6 | 1345 | ||
AK296458.1 | - | 6 | 1296 | |||
AK298278.1 | - | 7 | 811 | |||
AK303922.1 | - | 6 | 1117 | |||
AK312820.1 | - | 6 | 1357 | |||
BC105958.1 | - | 5 | 1236 | 40035034 | ||
BC107493.1 | - | 5 | 1302 | 40035036 | ||
BX537976.1 | - | 6 | 1356 |