Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TOR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000113.2 | NP_000104.1 | 5 | 851 | ||
GenBank mRNA | AF007871.1 | - | 5 | 816 | ||
AK056062.1 | - | 6 | 730 | |||
AK075343.1 | - | 5 | 801 | |||
AK130769.1 | - | 4 | 421 | |||
AK294677.1 | - | 5 | 941 | |||
AK314505.1 | - | 5 | 795 |