Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ELAVL2 SIRNA Assays ›
Gene Symbol:
ELAVL2
Gene Name:
ELAV like neuron-specific RNA binding protein 2
Gene Aliases:
HEL-N1, HELN1, HUB
Chromosome Location:
Chr.9: 23690099 - 23850757 on Build GRCh38
UniGene ID:
Hs.166109
Species:
Human
siRNA ID
HSS103172
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ELAVL2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001171195.1
NP_001164666.1
2
309
NM_001171197.1
NP_001164668.1
2
322
NM_004432.3
NP_004423.2
2
328
XM_005251393.3
XP_005251450.1
2
466
XM_005251394.3
XP_005251451.1
2
623
XM_005251395.3
XP_005251452.1
2
403
XM_006716734.3
XP_006716797.1
3
715
XM_006716735.3
XP_006716798.1
3
183
XM_006716736.3
XP_006716799.1
2
604
XM_011517774.2
XP_011516076.1
3
729
XM_011517776.2
XP_011516078.1
2
159
XM_011517777.2
XP_011516079.1
3
715
XM_011517778.2
XP_011516080.2
2
252
XM_011517779.2
XP_011516081.1
3
227
XM_011517780.2
XP_011516082.1
3
556
XM_011517783.2
XP_011516085.1
3
589
XM_011517784.2
XP_011516086.1
2
284
XM_011517785.2
XP_011516087.1
2
89
XM_011517786.2
XP_011516088.1
2
463
XM_017014408.1
XP_016869897.1
4
730
XM_017014409.1
XP_016869898.1
3
368
XM_017014410.1
XP_016869899.1
3
512
XM_017014411.1
XP_016869900.1
3
695
XM_017014412.1
XP_016869901.1
4
677
XM_017014413.1
XP_016869902.1
4
676
XM_017014414.1
XP_016869903.1
3
699
XM_017014415.1
XP_016869904.1
2
145
XM_017014416.1
XP_016869905.1
3
218
XM_017014417.1
XP_016869906.1
3
589
XM_017014418.1
XP_016869907.1
2
423
XM_017014419.1
XP_016869908.1
2
314
XM_017014420.1
XP_016869909.1
2
317
XM_017014421.1
XP_016869910.1
2
435
XM_017014422.1
XP_016869911.1
2
604
XM_017014423.1
XP_016869912.1
2
261
XM_017014424.1
XP_016869913.1
2
607
XM_017014425.1
XP_016869914.1
2
415
GenBank mRNA
AB209294.1
-
2
328
BC030692.1
-
2
326
4826082
BC042393.1
-
2
312
Back To Top
More Information
Panther Classification:
Molecular Function -
translation initiation factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
regulation of transcription, DNA-templated
nucleotide binding
mRNA 3'-UTR binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-ftf7j:80/10.239.160.32:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0