Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC29A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078175.2 | NP_001071643.1 | 5 | 524 | ||
NM_001078177.1 | NP_001071645.1 | 5 | 719 | |||
NM_001304462.1 | NP_001291391.1 | 5 | 657 | |||
NM_001304463.1 | NP_001291392.1 | 4 | 417 | |||
NM_001304465.1 | NP_001291394.1 | 4 | 560 | |||
NM_001304466.1 | NP_001291395.1 | 4 | 557 | |||
XM_005248876.4 | XP_005248933.1 | 4 | 382 | |||
XM_005248878.3 | XP_005248935.1 | 5 | 554 | |||
XM_005248879.3 | XP_005248936.1 | 5 | 488 | |||
XM_005248880.3 | XP_005248937.1 | 5 | 465 | |||
XM_005248881.3 | XP_005248938.1 | 4 | 562 | |||
XM_005248882.3 | XP_005248939.1 | 4 | 505 | |||
XM_011514341.2 | XP_011512643.1 | 5 | 578 | |||
GenBank mRNA | AF079117.1 | - | 4 | 330 | ||
AK090491.1 | - | 5 | 526 | |||
AK090615.1 | - | 5 | 545 | |||
AK293558.1 | - | 4 | 326 | |||
AK304285.1 | - | 3 | 346 | |||
AX775997.1 | - | 4 | 406 | |||
BC001382.2 | - | 4 | 330 | 3051441 | ||
BC008954.2 | - | 5 | 403 | 3010092 | ||
CB996332.1 | - | 5 | 496 | |||
DC396448.1 | - | 4 | 417 | |||
U81375.1 | - | 4 | 406 |