Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EPB41L2 SIRNA Assays ›
Gene Symbol:
EPB41L2
Gene Name:
erythrocyte membrane protein band 4.1 like 2
Gene Aliases:
4.1-G, 4.1G
Chromosome Location:
Chr.6: 130829111 - 131063322 on Build GRCh38
UniGene ID:
Hs.486470
Species:
Human
siRNA ID
HSS103271
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EPB41L2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001199388.2
NP_001186317.1
15
2525
NM_001252660.1
NP_001239589.1
14
2177
NM_001431.3
NP_001422.1
17
2971
XM_006715362.2
XP_006715425.1
15
2792
XM_011535524.1
XP_011533826.1
19
3127
XM_011535525.1
XP_011533827.1
18
3073
XM_011535527.1
XP_011533829.1
18
3025
XM_011535528.1
XP_011533830.1
16
2815
XM_011535529.1
XP_011533831.1
15
2761
XM_011535530.1
XP_011533832.1
15
2512
XM_011535534.1
XP_011533836.1
14
2197
XM_011535536.1
XP_011533838.1
13
2074
XM_017010349.1
XP_016865838.1
19
3155
XM_017010350.1
XP_016865839.1
19
3089
XM_017010351.1
XP_016865840.1
19
3151
XM_017010352.1
XP_016865841.1
19
2993
XM_017010353.1
XP_016865842.1
19
3127
XM_017010354.1
XP_016865843.1
18
3064
XM_017010355.1
XP_016865844.1
17
2999
XM_017010356.1
XP_016865845.1
19
2878
XM_017010357.1
XP_016865846.1
15
2723
XM_017010358.1
XP_016865847.1
16
2566
XM_017010359.1
XP_016865848.1
15
2540
XM_017010360.1
XP_016865849.1
15
2378
XM_017010361.1
XP_016865850.1
14
2225
XM_017010362.1
XP_016865851.1
14
2159
XM_017010363.1
XP_016865852.1
13
2064
XM_017010364.1
XP_016865853.1
17
2376
GenBank mRNA
AF027299.1
-
17
2833
AF054999.1
-
3
221
AK295958.1
-
7
984
AL050384.1
-
2
1516
AY047584.1
-
16
2797
AY512660.1
-
6
2392
CR749262.1
-
14
2139
CR936793.1
-
14
2177
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cortical actin cytoskeleton organization
actin binding
structural molecule activity
spectrin binding
PH domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0