Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000123.3 | NP_000114.2 | 5, 6 | 956 | ||
GenBank mRNA | AF462447.1 | - | 5, 6 | 913 | ||
AK294708.1 | - | 5, 6 | 885 | |||
AK299758.1 | - | 5, 6 | 668 | |||
AK303558.1 | - | 6, 7 | 859 | |||
BC031522.2 | - | 5, 6 | 942 | 3869893 | ||
BX647399.1 | - | 5, 6 | 785 | |||
D16305.1 | - | 5, 6 | 540 | |||
L20046.1 | - | 5, 6 | 578 | |||
LM994821.1 | - | 1, 2 | 26 | |||
X69978.1 | - | 5, 6 | 727 |
Gene Symbol: BIVM-ERCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204425.1 | NP_001191354.1 | 13, 14 | 2014 | ||
GenBank mRNA | AB209312.1 | - | 13, 14 | 1806 | ||
LM994822.1 | - | 13, 14 | 1892 |