Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: EVI2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003927.2 | NP_001003927.1 | 3 | 630 | ||
NM_014210.3 | NP_055025.2 | 2 | 489 | |||
GenBank mRNA | AK295311.1 | - | 3 | 565 | ||
AK312348.1 | - | 2 | 442 | |||
AV724051.1 | - | 2 | 410 | |||
BC035572.1 | - | 2 | 377 | 5229926 | ||
BG699778.1 | - | 3 | 530 | |||
BG700097.1 | - | 3 | 587 | |||
BI597493.1 | - | 2 | 445 | |||
DA364537.1 | - | 2 | 489 |