Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000137.2 | NP_000128.1 | 8 | 853 | ||
GenBank mRNA | AK313951.1 | - | 8 | 848 | ||
BC002527.1 | - | 8 | 691 | 3139988 | ||
BT007160.1 | - | 8 | 689 | |||
BX537608.1 | - | 7 | 2718 | |||
M55150.1 | - | 8 | 745 | |||
X51728.1 | - | 7 | 616 |