Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278113.1 | NP_001265042.1 | 16 | 1893 | ||
NM_001278114.1 | NP_001265043.1 | 16 | 1761 | |||
NM_001464.4 | NP_001455.3 | 17 | 1950 | |||
XM_005273468.1 | XP_005273525.1 | 17 | 1798 | |||
XM_011544479.1 | XP_011542781.1 | 13 | 1510 | |||
XM_011544480.1 | XP_011542782.1 | 13 | 1420 | |||
GenBank mRNA | AJ133005.1 | - | 17 | 1850 | ||
AK301734.1 | - | 16 | 1699 | |||
BC034957.2 | - | 16 | 1813 | 4826530 | ||
BC064547.1 | - | 13 | 1403 | 5744846 | ||
U38805.1 | - | 17 | 1914 | |||
U52370.1 | - | 17 | 1853 | |||
X99374.1 | - | 17 | 1846 |