Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319053.1 | NP_001305982.1 | 8 | 1114 | ||
NM_001470.3 | NP_001461.1 | 8 | 1178 | |||
NM_021903.2 | NP_068703.1 | 3 | 978 | |||
NM_021904.3 | NP_068704.2 | 7 | 992 | |||
XM_005248982.2 | XP_005249039.1 | 8 | 993 | |||
XM_006715047.3 | XP_006715110.1 | 7 | 793 | |||
XM_011514453.2 | XP_011512755.1 | 6 | 772 | |||
XM_011514455.1 | XP_011512757.1 | 8 | 993 | |||
XM_017010676.1 | XP_016866165.1 | 4 | 524 | |||
GenBank mRNA | AB208790.1 | - | 8 | 913 | ||
AF099148.1 | - | 8 | 1000 | |||
AF301005.1 | - | 7 | 895 | |||
AJ012185.1 | - | 7 | 892 | |||
AJ012186.1 | - | 3 | 541 | |||
AJ012187.1 | - | 6 | 706 | |||
AJ012288.1 | - | 8 | 935 | |||
AJ225028.1 | - | 8 | 1126 | |||
AJ225029.1 | - | 3 | 720 | |||
AK223619.1 | - | 3 | 763 | |||
AK290191.1 | - | 3 | 763 | |||
AK312308.1 | - | 3 | 759 | |||
BC041332.2 | - | 3 | 763 | 5275467 | ||
BC042598.1 | - | 3 | 752 | 4938491 | ||
BC050532.1 | - | 3 | 750 | 5732186 | ||
Y11044.1 | - | 7 | 889 |
Set Membership: |
![]() |