Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319053.1 | NP_001305982.1 | 9 | 1249 | ||
NM_001470.3 | NP_001461.1 | 9 | 1313 | |||
NM_021903.2 | NP_068703.1 | 4 | 1113 | |||
NM_021904.3 | NP_068704.2 | 8 | 1127 | |||
XM_005248982.2 | XP_005249039.1 | 9 | 1128 | |||
XM_006715047.3 | XP_006715110.1 | 8 | 928 | |||
XM_011514453.2 | XP_011512755.1 | 7 | 907 | |||
XM_011514455.1 | XP_011512757.1 | 9 | 1128 | |||
XM_017010676.1 | XP_016866165.1 | 5 | 659 | |||
GenBank mRNA | AB208790.1 | - | 9 | 1048 | ||
AF099148.1 | - | 9 | 1135 | |||
AF301005.1 | - | 8 | 1030 | |||
AJ012185.1 | - | 8 | 1027 | |||
AJ012186.1 | - | 4 | 676 | |||
AJ012187.1 | - | 7 | 841 | |||
AJ012288.1 | - | 9 | 1070 | |||
AJ225028.1 | - | 9 | 1261 | |||
AJ225029.1 | - | 4 | 855 | |||
AK223619.1 | - | 4 | 898 | |||
AK290191.1 | - | 4 | 898 | |||
BC041332.2 | - | 4 | 898 | 5275467 | ||
BC042598.1 | - | 4 | 887 | 4938491 | ||
BC050532.1 | - | 4 | 885 | 5732186 | ||
Y11044.1 | - | 8 | 1024 |
Set Membership: |
![]() |