Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128922.1 | NP_001122394.1 | 3 | 1142 | ||
NM_005512.2 | NP_005503.1 | 3 | 1247 | |||
XM_005273902.3 | XP_005273959.1 | 3 | 1336 | |||
XM_005273903.4 | XP_005273960.2 | 3 | 1229 | |||
XM_011544935.2 | XP_011543237.1 | 3 | 1538 | |||
XM_017017527.1 | XP_016873016.1 | 3 | 1742 | |||
GenBank mRNA | AK314971.1 | - | 3 | 1221 | ||
BC052210.1 | - | 3 | 1139 | 4825609 | ||
BC070079.1 | - | 3 | 1142 | 30344529 | ||
Z24680.1 | - | 3 | 1098 |