Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GNAS SIRNA Assays ›
Gene Symbol:
GNAS
Gene Name:
GNAS complex locus
Gene Aliases:
AHO, C20orf45, GNAS1, GPSA, GSA, GSP, NESP, POH, SCG6, SgVI
Chromosome Location:
Chr.20: 58839681 - 58911196 on Build GRCh38
UniGene ID:
Hs.125898
Species:
Human
siRNA ID
HSS104241
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GNAS
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000516.5
NP_000507.1
10
1192
NM_001077488.3
NP_001070956.1
11
1195
NM_001077489.3
NP_001070957.1
9
1147
NM_001077490.2
NP_001070958.1
10
2982
NM_001309840.1
NP_001296769.1
10
858
NM_001309861.1
NP_001296790.1
10
903
NM_016592.3
NP_057676.1
11
1801
NM_080425.3
NP_536350.2
10
2982
NM_080426.3
NP_536351.1
10
1150
XM_017027812.1
XP_016883301.1
11
3252
XM_017027813.1
XP_016883302.1
10
3207
XM_017027814.1
XP_016883303.1
9
3204
XM_017027815.1
XP_016883304.1
9
675
XM_017027816.1
XP_016883305.1
11
1480
XM_017027817.1
XP_016883306.1
9
762
XM_017027818.1
XP_016883307.1
9
685
XM_017027819.1
XP_016883308.1
9
726
XM_017027820.1
XP_016883309.1
9
666
GenBank mRNA
AF105253.1
-
11
1779
AF493897.1
-
10
768
AF493898.1
-
10
726
AK026564.1
-
11
962
AK054862.1
-
8
2056
AK093534.1
-
9
1002
AK122771.1
-
8
1175
AK225818.1
-
10
891
AK315860.1
-
10
690
AK315874.1
-
10
871
AL537118.3
-
10
888
BC002722.2
-
11
886
BC008855.2
-
12
913
BC022875.1
-
9
614
BC066923.1
-
9
726
BC089157.1
-
10
1042
BC104928.1
-
10
814
BC108315.1
-
9
988
BT009905.1
-
10
768
KU178095.1
-
10
768
KU178096.1
-
10
726
M14631.1
-
11
738
X04408.1
-
10
836
X04409.1
-
10
738
X07036.1
-
10
780
X56009.1
-
10
844
Back To Top
More Information
Panther Classification:
Molecular Function -
heterotrimeric G-protein
Gene Ontology Categories:
Function(s)
Process(es)
tissue homeostasis
endochondral ossification
energy reserve metabolic process
DNA methylation
adenylate cyclase-activating G-protein coupled receptor signaling pathway
adenylate cyclase-activating dopamine receptor signaling pathway
sensory perception of chemical stimulus
embryonic hindlimb morphogenesis
multicellular organism growth
post-embryonic body morphogenesis
response to drug
positive regulation of osteoblast differentiation
positive regulation of osteoclast differentiation
developmental growth
embryonic cranial skeleton morphogenesis
cognition
cartilage development
bone development
hair follicle placode formation
platelet aggregation
genetic imprinting
regulation of parathyroid hormone secretion
GTPase activity
signal transducer activity
insulin-like growth factor receptor binding
protein binding
GTP binding
G-protein beta/gamma-subunit complex binding
beta-2 adrenergic receptor binding
D1 dopamine receptor binding
mu-type opioid receptor binding
ionotropic glutamate receptor binding
metal ion binding
corticotropin-releasing hormone receptor 1 binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0