Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002079.2 | NP_002070.1 | 8, 9 | 1320 | ||
GenBank mRNA | AF052153.1 | - | 8, 9 | 1140 | ||
AK293483.1 | - | 8, 9 | 1042 | |||
AK301916.1 | - | 8, 9 | 1218 | |||
AK312684.1 | - | 8, 9 | 1162 | |||
AY513279.1 | - | 8, 9 | 1114 | |||
BC000498.2 | - | 8, 9 | 1140 | 2820849 | ||
M37400.1 | - | 8, 9 | 1136 |