Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRLHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004248.2 | NP_004239.1 | 2 | 736 | ||
GenBank mRNA | AB015745.1 | - | 1 | 597 | ||
AK290461.1 | - | 2 | 736 | |||
AK313269.1 | - | 2 | 738 | |||
BC095539.1 | - | 1 | 722 | 30706566 | ||
BC101490.1 | - | 1 | 671 | 8068996 | ||
BC101492.1 | - | 1 | 671 | 8068998 | ||
EF577402.1 | - | 1 | 597 |
Set Membership: |
G-protein coupled receptor |