Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GRK4 SIRNA Assays ›
Gene Symbol:
GRK4
Gene Name:
G protein-coupled receptor kinase 4
Gene Aliases:
GPRK2L, GPRK4, GRK4a, IT11
Chromosome Location:
Chr.4: 2963504 - 3040760 on Build GRCh38
UniGene ID:
Hs.32959
Species:
Human
siRNA ID
HSS104406
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GRK4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001004056.1
NP_001004056.1
12, 13
1777
NM_001004057.1
NP_001004057.1
13, 14
1873
NM_005307.2
NP_005298.2
12, 13
1777
NM_182982.2
NP_892027.2
13, 14
1873
XM_005247962.3
XP_005248019.1
11, 12
1391
XM_006713880.3
XP_006713943.1
12, 13
1289
XM_011513447.2
XP_011511749.1
14, 15
2102
XM_011513448.2
XP_011511750.1
14, 15
2102
XM_011513449.2
XP_011511751.1
13, 14
2006
XM_011513450.2
XP_011511752.1
14, 15
2102
XM_011513451.2
XP_011511753.1
13, 14
1964
XM_011513452.2
XP_011511754.1
13, 14
1964
XM_011513454.2
XP_011511756.1
13, 14
1964
XM_017008052.1
XP_016863541.1
13, 14
2024
XM_017008053.1
XP_016863542.1
13, 14
2012
XM_017008054.1
XP_016863543.1
12, 13
1847
XM_017008055.1
XP_016863544.1
11, 12
1751
XM_017008057.1
XP_016863546.1
10, 11
1189
XM_017008058.1
XP_016863547.1
12, 13
1249
XM_017008059.1
XP_016863548.1
10, 11
1273
XM_017008060.1
XP_016863549.1
11, 12
1367
XM_017008061.1
XP_016863550.1
12, 13
1640
XM_017008062.1
XP_016863551.1
12, 13
1787
XM_017008063.1
XP_016863552.1
11, 12
1576
XM_017008064.1
XP_016863553.1
10, 11
1222
XM_017008066.1
XP_016863555.1
9, 10
1059
GenBank mRNA
AK223581.1
-
12, 13
1785
AK292390.1
-
13, 14
1918
BC117320.1
-
13, 14
1761
40125871
BM725978.1
-
1, 2
139
U33054.1
-
13, 14
1672
U33055.1
-
12, 13
1576
U33056.1
-
13, 14
1672
X97879.1
-
12, 13
1420
X97880.1
-
12, 13
1420
X97881.1
-
13, 14
1516
X98118.1
-
13, 14
1516
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor internalization
protein phosphorylation
signal transduction
regulation of G-protein coupled receptor protein signaling pathway
regulation of rhodopsin mediated signaling pathway
receptor internalization
G-protein coupled receptor kinase activity
ATP binding
rhodopsin kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-wh4bk:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0