Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005513.2 | NP_005504.2 | 3 | 721 | ||
XM_011512744.2 | XP_011511046.1 | 3 | 844 | |||
XM_011512745.2 | XP_011511047.1 | 3 | 792 | |||
GenBank mRNA | S67859.1 | - | 3 | 670 | ||
X63468.1 | - | 2 | 661 |