Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2H4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001517.4 | NP_001508.1 | 2 | 239 | ||
GenBank mRNA | AK222607.1 | - | 2 | 143 | ||
AK298058.1 | - | 2 | 287 | |||
AK309869.1 | - | 2 | 253 | |||
BC004935.1 | - | 2 | 211 | 3606718 | ||
BC016302.2 | - | 2 | 211 | 3830902 | ||
BT007321.1 | - | 1 | 32 | |||
Y07595.1 | - | 2 | 159 |