Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFHR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001312672.1 | NP_001299601.1 | 3 | 530 | ||
NM_005666.3 | NP_005657.1 | 5 | 902 | |||
XM_005245113.3 | XP_005245170.1 | 4 | 721 | |||
XM_011509458.2 | XP_011507760.1 | 5 | 874 | |||
XM_011509459.2 | XP_011507761.1 | 5 | 936 | |||
XM_011509460.2 | XP_011507762.1 | 4 | 711 | |||
XM_017001109.1 | XP_016856598.1 | 4 | 709 | |||
GenBank mRNA | AX772812.1 | - | 3 | 440 | ||
BC022283.1 | - | 5 | 891 | 4734331 | ||
BG566607.1 | - | 3 | 518 | |||
X64877.1 | - | 5 | 866 |