Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HRH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000861.3 | NP_000852.1 | 3 | 1641 | ||
NM_001098211.1 | NP_001091681.1 | 2 | 1562 | |||
NM_001098212.1 | NP_001091682.1 | 2 | 1512 | |||
NM_001098213.1 | NP_001091683.1 | 2 | 1792 | |||
XM_011533652.1 | XP_011531954.1 | 4 | 1929 | |||
XM_011533653.2 | XP_011531955.1 | 2 | 1758 | |||
XM_017006283.1 | XP_016861772.1 | 2 | 1712 | |||
XM_017006284.1 | XP_016861773.1 | 2 | 1751 | |||
GenBank mRNA | AF026261.1 | - | 1 | 1450 | ||
AK289412.1 | - | 2 | 1792 | |||
AK292157.1 | - | 2 | 1509 | |||
AK312879.1 | - | 2 | 1510 | |||
AY136743.1 | - | 1 | 1450 | |||
BC060802.1 | - | 3 | 1641 | 30340368 | ||
D28481.1 | - | 1 | 1453 | |||
Z34897.1 | - | 1 | 1450 |
Set Membership: |
![]() |