Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000414.3 | NP_000405.1 | 15 | 1422 | ||
NM_001199291.2 | NP_001186220.1 | 16 | 1621 | |||
NM_001199292.1 | NP_001186221.1 | 14 | 1368 | |||
NM_001292027.1 | NP_001278956.1 | 16 | 1487 | |||
NM_001292028.1 | NP_001278957.1 | 15 | 1547 | |||
GenBank mRNA | AK094049.1 | - | 15 | 1291 | ||
AK293254.1 | - | 15 | 1294 | |||
AK295440.1 | - | 10 | 1034 | |||
AK298075.1 | - | 16 | 1549 | |||
AK299685.1 | - | 16 | 1392 | |||
AK301212.1 | - | 14 | 1314 | |||
AK308968.1 | - | 11 | 1136 | |||
AK312449.1 | - | 15 | 1368 | |||
BC003098.1 | - | 15 | 1356 | 3502915 | ||
KU178135.1 | - | 15 | 1289 | |||
X87176.1 | - | 15 | 1337 |