Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ICA1 SIRNA Assays ›
Gene Symbol:
ICA1
Gene Name:
islet cell autoantigen 1
Gene Aliases:
ICA69, ICAp69
Chromosome Location:
Chr.7: 8113184 - 8262687 on Build GRCh38
UniGene ID:
Hs.487561
Species:
Human
siRNA ID
HSS105157
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ICA1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136020.2
NP_001129492.1
3
370
NM_001276478.1
NP_001263407.1
3
352
NM_004968.3
NP_004959.2
3
355
NM_022307.2
NP_071682.1
3
170
XM_005249735.3
XP_005249792.1
3
292
XM_011515345.1
XP_011513647.1
3
370
XM_011515346.1
XP_011513648.1
3
355
XM_011515347.2
XP_011513649.1
3
242
XM_011515348.1
XP_011513650.1
3
288
XM_011515349.1
XP_011513651.1
3
221
XM_011515350.1
XP_011513652.1
3
292
XM_011515351.1
XP_011513653.1
3
427
XM_011515352.1
XP_011513654.1
3
246
XM_011515353.2
XP_011513655.1
3
299
XM_017012116.1
XP_016867605.1
3
138
XM_017012117.1
XP_016867606.1
3
328
XM_017012118.1
XP_016867607.1
3
288
XM_017012119.1
XP_016867608.1
3
263
XM_017012120.1
XP_016867609.1
3
309
XM_017012121.1
XP_016867610.1
3
176
XM_017012122.1
XP_016867611.1
3
341
XM_017012123.1
XP_016867612.1
3
280
XM_017012124.1
XP_016867613.1
3
441
XM_017012125.1
XP_016867614.1
3
191
GenBank mRNA
AK292106.1
-
3
239
BC008640.2
-
3
287
3854722
DA076379.1
-
3
356
DA154134.1
-
3
370
DB010744.1
-
3
339
DB040266.1
-
3
298
EU416199.1
-
2
103
L01100.1
-
3
281
L21181.1
-
2
179
U26591.1
-
3
170
U26592.1
-
3
227
U37183.1
-
1
69
U38260.1
-
3
272
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neurotransmitter transport
regulation of protein homodimerization activity
regulation of insulin secretion
molecular_function
protein binding
protein domain specific binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qvdz4:80/10.239.165.207:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0