Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006083.3 | NP_006074.2 | 10 | 959 | ||
GenBank mRNA | AF182645.1 | - | 10 | 960 | ||
AJ005579.1 | - | 10 | 849 | |||
AK225924.1 | - | 10 | 943 | |||
AK293626.1 | - | 10 | 787 | |||
AK308730.1 | - | 10 | 797 | |||
AK312363.1 | - | 10 | 929 | |||
AY007094.1 | - | 10 | 882 | |||
BC013005.2 | - | 10 | 914 | 3030079 | ||
BC051295.1 | - | 10 | 917 | |||
BC071964.1 | - | 10 | 907 | 6461871 |