Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: IL1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000576.2 | NP_000567.1 | 3 | 154 | ||
XM_017003988.1 | XP_016859477.1 | 2 | 155 | |||
GenBank mRNA | AK313983.1 | - | 2 | 617 | ||
BC008678.1 | - | 3 | 154 | 3875593 | ||
BT007213.1 | - | 2 | 67 | |||
CR407679.1 | - | 2 | 67 | |||
K02770.1 | - | 3 | 153 | |||
M15330.1 | - | 3 | 153 | |||
X02532.1 | - | 3 | 144 | |||
X56087.1 | - | 3 | 124 |