Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IREB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320941.1 | NP_001307870.1 | 14 | 1982 | ||
NM_001320942.1 | NP_001307871.1 | 15 | 2306 | |||
NM_004136.3 | NP_004127.1 | 15 | 2052 | |||
GenBank mRNA | AB209403.1 | - | 15 | 1986 | ||
AK027033.1 | - | 14 | 1816 | |||
AK027034.1 | - | 6 | 670 | |||
AK292992.1 | - | 15 | 1936 | |||
BC117481.1 | - | 15 | 1919 | 40126032 | ||
BC117483.1 | - | 15 | 1919 | 40126034 | ||
BC143948.1 | - | 14 | 1849 | |||
M58511.1 | - | 15 | 1890 | |||
U20180.1 | - | 14 | 1857 |