Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITIH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166434.2 | NP_001159906.1 | 9 | 1243 | ||
NM_001166435.2 | NP_001159907.1 | 7 | 974 | |||
NM_001166436.2 | NP_001159908.1 | 7 | 860 | |||
NM_002215.3 | NP_002206.2 | 11 | 1404 | |||
GenBank mRNA | AF116713.2 | - | 2 | 266 | ||
AK292750.1 | - | 11 | 1374 | |||
AK298398.1 | - | 11 | 1403 | |||
AK298423.1 | - | 11 | 1396 | |||
AK298455.1 | - | 9 | 1229 | |||
AK303110.1 | - | 7 | 860 | |||
AK303156.1 | - | 7 | 974 | |||
AK314198.1 | - | 11 | 1374 | |||
BC069464.1 | - | 11 | 1398 | 7262464 | ||
M18192.1 | - | 2 | 181 | |||
X16260.1 | - | 9 | 1153 | |||
X63652.1 | - | 11 | 1398 |