Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099952.2 | NP_001093422.2 | 57 | 8213 | ||
NM_001168272.1 | NP_001161744.1 | 59 | 8312 | |||
NM_002222.5 | NP_002213.5 | 56 | 8168 | |||
XM_005265109.2 | XP_005265166.1 | 58 | 8288 | |||
XM_005265110.2 | XP_005265167.1 | 57 | 8240 | |||
XM_006713131.2 | XP_006713194.1 | 58 | 8291 | |||
XM_011533681.1 | XP_011531983.1 | 60 | 8360 | |||
XM_011533682.2 | XP_011531984.1 | 60 | 8286 | |||
XM_011533683.2 | XP_011531985.1 | 60 | 8357 | |||
XM_011533684.1 | XP_011531986.1 | 60 | 8333 | |||
XM_011533685.1 | XP_011531987.1 | 59 | 8327 | |||
XM_011533686.1 | XP_011531988.1 | 59 | 8324 | |||
XM_011533687.1 | XP_011531989.1 | 59 | 8315 | |||
XM_011533688.1 | XP_011531990.1 | 59 | 8288 | |||
XM_011533690.1 | XP_011531992.1 | 59 | 8237 | |||
XM_017006357.1 | XP_016861846.1 | 59 | 8234 | |||
GenBank mRNA | AB208868.1 | - | 37 | 5084 | ||
AK294594.1 | - | 8 | 1285 | |||
AK296235.1 | - | 8 | 1309 | |||
AK309981.1 | - | 8 | 1264 | |||
BC144563.1 | - | 18 | 2044 | |||
D26070.1 | - | 56 | 8074 | |||
L38019.2 | - | 57 | 8110 | |||
U23850.1 | - | 57 | 7962 |