Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004984.2 | NP_004975.2 | 14 | 1628 | ||
GenBank mRNA | AB210045.1 | - | 14 | 1453 | ||
AB290174.1 | - | 14 | 1694 | |||
AK294881.1 | - | 11 | 1381 | |||
AK315202.1 | - | 14 | 1648 | |||
BC146670.1 | - | 14 | 1631 | 40148192 | ||
BC150208.1 | - | 14 | 1631 | 40148890 | ||
U06698.1 | - | 14 | 1568 |