Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002272.3 | NP_002263.3 | 7 | 1199 | ||
GenBank mRNA | AK056254.1 | - | 7 | 1676 | ||
AK098110.1 | - | 7 | 1227 | |||
AK296582.1 | - | 8 | 1166 | |||
AK299394.1 | - | 8 | 1142 | |||
AK299400.1 | - | 7 | 1199 | |||
AK299451.1 | - | 8 | 1061 | |||
AK307941.1 | - | 7 | 945 | |||
BC003630.2 | - | 6 | 555 | |||
BC042174.1 | - | 1204 | 5453644 | |||
X07695.1 | - | 7 | 805 |