Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000226.3 | NP_000217.2 | 4 | 1042 | ||
GenBank mRNA | AB001594.1 | - | 4 | 539 | ||
BC121170.2 | - | 5 | 508 | |||
S69510.1 | - | 4 | 1039 | |||
Z29074.1 | - | 4 | 1042 |