Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160102.1 | NP_001153574.1 | 6 | 882 | ||
NM_006151.2 | NP_006142.1 | 8 | 1131 | |||
NR_027647.1 | 7 | 1043 | ||||
XM_011524808.2 | XP_011523110.1 | 8 | 1007 | |||
XM_011524810.2 | XP_011523112.1 | 2 | 606 | |||
GenBank mRNA | AK303903.1 | - | 7 | 878 | ||
AK308378.1 | - | 8 | 856 | |||
BC107166.1 | - | 8 | 1002 | 40034365 | ||
BC107167.1 | - | 8 | 1002 | 40034371 | ||
BX648309.1 | - | 8 | 969 | |||
EF579964.1 | - | 6 | 604 | |||
U39573.1 | - | 8 | 896 |