Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: M6PR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207024.1 | NP_001193953.1 | 5 | 768 | ||
NM_002355.3 | NP_002346.1 | 7 | 1026 | |||
XM_005253376.2 | XP_005253433.1 | 7 | 888 | |||
GenBank mRNA | AK222792.1 | - | 7 | 907 | ||
AK291143.1 | - | 7 | 872 | |||
AK312689.1 | - | 7 | 911 | |||
BC017111.1 | - | 5 | 426 | |||
BC024206.1 | - | 7 | 918 | 3530887 | ||
BM546062.1 | - | 2 | 181 | |||
BU166950.1 | - | 5 | 663 | |||
CR457412.1 | - | 6 | 751 | |||
M16985.1 | - | 7 | 896 |