Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN2A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002372.3 | NP_002363.2 | 19 | 3972 | ||
XM_017009472.1 | XP_016864961.1 | 19 | 2893 | |||
GenBank mRNA | AK314090.1 | - | 19 | 3313 | ||
BC142656.1 | - | 19 | 3274 | 40148064 | ||
BC142696.1 | - | 19 | 3274 | 40148740 | ||
D63998.1 | - | 19 | 3430 | |||
U31520.1 | - | 19 | 2923 |