Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN2B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000528.3 | NP_000519.2 | 22 | 2841 | ||
NM_001173498.1 | NP_001166969.1 | 22 | 2838 | |||
XM_005259913.1 | XP_005259970.1 | 22 | 2822 | |||
XM_017026818.1 | XP_016882307.1 | 19 | 2420 | |||
GenBank mRNA | AB209921.1 | - | 22 | 2753 | ||
AK291572.1 | - | 22 | 2806 | |||
AK303418.1 | - | 23 | 2780 | |||
BC000736.2 | - | 22 | 2787 | 3504417 | ||
BM982008.1 | - | 3 | 398 | |||
U05572.1 | - | 22 | 2795 | |||
U60266.1 | - | 22 | 2779 | |||
U68382.1 | - | 20 | 3354 | |||
U68567.1 | - | 22 | 2753 |