Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324255.1 | NP_001311184.1 | 4 | 6055 | ||
NM_005909.4 | NP_005900.2 | 5 | 6325 | |||
GenBank mRNA | CR933720.1 | - | 1 | 3973 |