Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000242.2 | NP_000233.1 | 1 | 111 | ||
XM_006717861.3 | XP_006717924.1 | 2 | 179 | |||
XM_011539816.2 | XP_011538118.1 | 2 | 163 | |||
GenBank mRNA | AF360991.1 | - | 1 | 62 | ||
AY826184.1 | - | 1 | 46 | |||
BC069338.1 | - | 1 | 96 | 7262231 | ||
BC096179.3 | - | 1 | 77 | 40003735 | ||
BC096180.3 | - | 1 | 77 | 40003741 | ||
BC096181.3 | - | 1 | 77 | 40003742 | ||
BC096182.3 | - | 1 | 77 | 40003743 | ||
X15422.1 | - | 1 | 111 |