Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MEF2A SIRNA Assays ›
Gene Symbol:
MEF2A
Gene Name:
myocyte enhancer factor 2A
Gene Aliases:
ADCAD1, RSRFC4, RSRFC9, mef2
Chromosome Location:
Chr.15: 99565455 - 99716488 on Build GRCh38
UniGene ID:
Hs.268675
Species:
Human
siRNA ID
HSS106436
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MEF2A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130926.2
NP_001124398.1
10
1449
NM_001130927.2
NP_001124399.1
10
1269
NM_001130928.2
NP_001124400.1
8
1068
NM_001171894.2
NP_001165365.1
12
1739
NM_001319206.1
NP_001306135.1
12
1555
NM_005587.3
NP_005578.2
11
1537
XM_005254914.2
XP_005254971.1
11
1514
XM_005254915.2
XP_005254972.1
11
1759
XM_011521571.2
XP_011519873.1
12
1538
XM_011521572.2
XP_011519874.1
12
1483
XM_011521573.2
XP_011519875.1
12
1513
XM_011521576.2
XP_011519878.1
11
1427
XM_011521577.2
XP_011519879.1
11
1361
XM_011521578.2
XP_011519880.1
11
2020
XM_011521579.2
XP_011519881.1
11
1437
XM_011521581.2
XP_011519883.1
12
1789
XM_011521582.2
XP_011519884.1
13
1994
XM_011521583.2
XP_011519885.1
11
1409
XM_011521585.2
XP_011519887.1
11
1597
XM_011521586.2
XP_011519888.1
11
1579
XM_011521587.2
XP_011519889.1
10
1555
XM_017022190.1
XP_016877679.1
11
1415
XM_017022191.1
XP_016877680.1
11
1407
XM_017022192.1
XP_016877681.1
10
1392
XM_017022193.1
XP_016877682.1
11
1575
XM_017022194.1
XP_016877683.1
10
1391
XM_017022195.1
XP_016877684.1
10
1248
XM_017022196.1
XP_016877685.1
13
1927
XM_017022197.1
XP_016877686.1
12
3706
XM_017022198.1
XP_016877687.1
13
1915
XM_017022199.1
XP_016877688.1
10
2192
XM_017022200.1
XP_016877689.1
11
1962
XM_017022201.1
XP_016877690.1
10
1585
XM_017022202.1
XP_016877691.1
13
1909
GenBank mRNA
AB208985.1
-
12
1716
AK291835.1
-
10
1359
AK294207.1
-
10
1207
AL831995.1
-
12
1416
BC013437.2
-
11
1455
3896491
BC053871.2
-
8
1040
5555678
KU178181.1
-
9
1136
KU178182.1
-
10
1160
KU178183.1
-
10
1154
KU178184.1
-
9
1130
KU178185.1
-
8
926
X63381.1
-
10
1364
X68505.1
-
12
1574
Y16312.1
-
10
1358
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mitochondrial genome maintenance
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
transcription, DNA-templated
transcription from RNA polymerase II promoter
apoptotic process
heart development
muscle organ development
positive regulation of cardiac muscle hypertrophy
positive regulation of transcription from RNA polymerase II promoter
mitochondrion distribution
dendrite morphogenesis
positive regulation of muscle cell differentiation
ventricular cardiac myofibril assembly
cardiac conduction
ERK5 cascade
cellular response to calcium ion
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
RNA polymerase II transcription factor binding
RNA polymerase II transcription coactivator activity
chromatin binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
protein binding
protein kinase binding
activating transcription factor binding
histone acetyltransferase binding
histone deacetylase binding
sequence-specific DNA binding
SMAD binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-j4kkz:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0