Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MIPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005932.3 | NP_005923.2 | 11 | 1288 | ||
XM_011535097.2 | XP_011533399.1 | 11 | 1051 | |||
XM_011535098.2 | XP_011533400.1 | 11 | 1288 | |||
GenBank mRNA | AK225290.1 | - | 11 | 1238 | ||
AK291923.1 | - | 11 | 1164 | |||
BC009934.2 | - | 11 | 1260 | 4053687 | ||
U80034.1 | - | 11 | 1234 |