Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATXN3 SIRNA Assays ›
Gene Symbol:
ATXN3
Gene Name:
ataxin 3
Gene Aliases:
AT3, ATX3, JOS, MJD, MJD1, SCA3
Chromosome Location:
Chr.14: 92058552 - 92106621 on Build GRCh38
UniGene ID:
Hs.532632
Species:
Human
siRNA ID
HSS106557
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATXN3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001127696.1
NP_001121168.1
7
658
NM_001127697.2
NP_001121169.2
6
550
NM_001164779.1
NP_001158251.1
4
340
NM_001164780.1
NP_001158252.1
5
407
NM_001164781.1
NP_001158253.1
6
493
NM_004993.5
NP_004984.2
8
703
NM_030660.4
NP_109376.1
7
538
NR_028453.1
7
617
NR_028454.1
6
452
NR_028455.1
7
617
NR_028456.1
6
452
NR_028457.1
8
708
NR_028458.1
7
594
NR_028459.1
8
703
NR_028461.1
7
615
NR_028462.1
7
594
NR_028463.1
3
252
NR_028464.1
6
550
NR_028465.1
6
572
NR_028466.1
3
252
NR_028467.1
7
618
NR_028469.1
6
452
NR_031765.1
2
119
GenBank mRNA
AB050194.1
-
7
527
AK225884.1
-
8
642
AK314919.1
-
8
655
BC022245.1
-
7
586
BC033711.1
-
8
646
4393766
BC095402.1
-
8
663
30347215
BU190081.1
-
7
637
BU663113.1
-
5
457
GQ176433.1
-
8
656
GQ176434.1
-
8
655
GQ176435.1
-
8
656
GQ176436.1
-
6
503
GQ176437.1
-
6
503
GQ176438.1
-
7
570
GQ176453.1
-
7
546
GQ176454.1
-
8
656
GQ176455.1
-
7
491
GQ176456.1
-
6
405
GQ176457.1
-
6
503
GQ176458.1
-
7
570
GQ176459.1
-
4
293
GQ176462.1
-
8
656
GQ176463.1
-
7
570
GQ176464.1
-
7
570
GQ176465.1
-
3
205
GQ176466.1
-
6
405
GQ176469.1
-
6
503
GQ176470.1
-
6
525
GQ176474.1
-
6
503
GQ176475.1
-
6
405
GQ176476.1
-
8
656
GQ176477.1
-
7
570
GQ176479.1
-
7
570
GQ176481.1
-
7
570
GQ176483.1
-
7
570
GQ176484.1
-
7
570
GQ176485.1
-
6
502
GQ176486.1
-
7
611
GQ176487.1
-
6
503
GQ176488.1
-
8
656
GQ176489.1
-
7
611
GQ176490.1
-
6
537
GQ176492.1
-
7
570
GQ176493.1
-
6
502
GQ176496.1
-
6
405
GQ176497.1
-
7
570
GQ176498.1
-
7
491
GQ176499.1
-
8
656
GQ176503.1
-
6
404
GQ176504.1
-
8
656
GQ176505.1
-
6
405
GQ176506.1
-
6
405
GQ176517.1
-
3
205
GQ176518.1
-
3
205
GQ176519.1
-
7
569
GQ176525.1
-
6
405
GQ176529.1
-
7
567
GQ176531.1
-
8
656
GQ176532.1
-
7
656
GQ176533.1
-
8
656
GQ176534.1
-
7
567
GQ176538.1
-
8
656
GQ176540.1
-
7
544
GQ176542.1
-
4
292
GQ176543.1
-
6
405
GQ176544.1
-
6
405
GQ176545.1
-
4
293
GQ176546.1
-
7
570
GQ176548.1
-
8
656
GQ176550.1
-
8
656
GQ176551.1
-
8
654
GQ176552.1
-
8
655
GQ176554.1
-
6
405
GQ176556.1
-
6
405
GQ176559.1
-
6
405
GQ176560.1
-
8
656
GQ176563.1
-
8
656
GQ176565.1
-
8
656
GQ176566.1
-
7
491
GQ176567.1
-
8
656
GQ176568.1
-
8
656
GQ176569.1
-
7
567
GQ176570.1
-
8
656
GQ176571.1
-
6
405
GQ176572.1
-
7
570
GQ176573.1
-
7
491
GQ176576.1
-
7
568
GQ176579.1
-
7
547
GQ176580.1
-
8
656
GQ176608.1
-
6
503
GQ176609.1
-
656
GQ176610.1
-
7
570
GQ176611.1
-
7
570
GQ176613.1
-
3
182
GQ176614.1
-
5
360
GQ176615.1
-
656
GQ176616.1
-
8
656
GQ176617.1
-
7
570
GQ176618.1
-
482
GQ176619.1
-
8
656
GQ176620.1
-
7
570
GQ176621.1
-
8
656
GQ176622.1
-
8
656
GQ176623.1
-
6
405
GQ176624.1
-
7
589
GQ176625.1
-
293
GQ176627.1
-
4
293
GQ176630.1
-
7
570
GQ176631.1
-
6
503
GQ176632.1
-
8
656
GQ176633.1
-
4
293
GQ176636.1
-
5
477
GQ176654.1
-
7
570
GQ176664.1
-
7
570
GQ176665.1
-
7
491
GQ176666.1
-
5
360
GQ176670.1
-
5
458
GQ176673.1
-
8
656
GQ176674.1
-
8
656
GQ176675.1
-
7
491
GQ176676.1
-
6
405
GQ176677.1
-
8
656
GQ176678.1
-
7
570
GQ176679.1
-
7
491
GQ176680.1
-
8
656
GQ176681.1
-
5
415
GQ176682.1
-
7
570
GQ176684.1
-
8
656
GQ176685.1
-
8
655
GQ176687.1
-
8
656
GQ176688.1
-
6
502
GQ176689.1
-
5
360
GQ176690.1
-
7
570
GQ176691.1
-
6
405
GQ176692.1
-
8
656
GQ176695.1
-
6
503
GQ176696.1
-
7
570
GQ176697.1
-
8
656
GQ176700.1
-
7
491
GQ176701.1
-
8
656
GQ176702.1
-
5
360
GQ176703.1
-
8
656
GQ176704.1
-
8
656
GQ176705.1
-
4
293
GQ176706.1
-
7
570
GQ176710.1
-
4
293
GQ176711.1
-
5
360
GQ176712.1
-
7
611
GQ176714.1
-
8
656
GQ176715.1
-
8
578
GQ176716.1
-
6
405
GQ176717.1
-
570
GQ176719.1
-
656
GQ176720.1
-
6
405
GQ176721.1
-
8
656
GQ176722.1
-
7
570
GQ176723.1
-
5
360
GQ176724.1
-
6
503
GQ176725.1
-
8
656
GQ176726.1
-
6
405
GQ176727.1
-
4
293
GQ176728.1
-
8
656
GQ176732.1
-
405
GQ176734.1
-
503
GQ176741.1
-
656
GQ176743.1
-
6
405
GQ176744.1
-
7
611
GQ176745.1
-
656
GQ176746.1
-
8
656
GQ176747.1
-
656
GQ176748.1
-
6
405
GQ176749.1
-
8
656
GQ176750.1
-
7
491
GQ176751.1
-
405
GQ176752.1
-
743
GQ176753.1
-
5
360
GQ176754.1
-
7
491
GQ176755.1
-
7
570
GQ176756.1
-
8
656
GQ176757.1
-
6
503
GQ176758.1
-
6
405
GQ176759.1
-
8
656
GQ176760.1
-
7
570
GQ176761.1
-
5
338
GQ176762.1
-
656
GQ176763.1
-
8
656
GQ176764.1
-
405
GQ176765.1
-
6
405
GQ176766.1
-
8
656
GQ176767.1
-
7
570
GQ176768.1
-
6
405
GQ176769.1
-
5
458
GQ176771.1
-
8
656
GQ176772.1
-
6
503
GQ176773.1
-
6
503
GQ176774.1
-
6
405
GQ176778.1
-
5
360
GQ176779.1
-
6
503
GQ176780.1
-
6
405
GQ176781.1
-
6
503
GQ176782.1
-
6
503
GQ176784.1
-
6
503
GQ176785.1
-
5
338
GQ176786.1
-
6
503
GQ176788.1
-
6
405
GQ176789.1
-
6
503
GQ176790.1
-
6
502
GQ176791.1
-
8
656
GQ176805.1
-
8
632
GQ176806.1
-
8
655
GQ176807.1
-
7
570
GQ176809.1
-
5
458
GQ176811.1
-
8
657
GQ176812.1
-
6
503
GQ176813.1
-
7
491
GQ176814.1
-
6
402
GQ176815.1
-
6
504
GQ176819.1
-
7
570
GQ176820.1
-
8
656
GQ176821.1
-
6
404
GQ176822.1
-
7
491
GQ176823.1
-
7
547
GQ176824.1
-
8
656
GQ176826.1
-
8
633
GQ176827.1
-
8
656
GQ176828.1
-
6
405
GQ176829.1
-
8
656
GQ176830.1
-
627
GQ176831.1
-
656
GQ176832.1
-
6
405
GQ176833.1
-
7
570
GQ176834.1
-
569
GQ176835.1
-
7
490
GQ176836.1
-
7
491
GQ176837.1
-
8
656
GQ176839.1
-
405
GQ176842.1
-
570
GQ176844.1
-
6
418
U64820.1
-
8
691
U64821.1
-
8
654
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
nucleotide-excision repair
transcription, DNA-templated
regulation of transcription, DNA-templated
ubiquitin-dependent protein catabolic process
misfolded or incompletely synthesized protein catabolic process
chemical synaptic transmission
nervous system development
regulation of cell-substrate adhesion
protein deubiquitination
actin cytoskeleton organization
cellular response to heat
monoubiquitinated protein deubiquitination
exploration behavior
proteasome-mediated ubiquitin-dependent protein catabolic process
intermediate filament cytoskeleton organization
protein K63-linked deubiquitination
histone H3 deacetylation
protein K48-linked deubiquitination
cellular response to misfolded protein
positive regulation of ERAD pathway
protein localization to cytosolic proteasome complex involved in ERAD pathway
RNA polymerase II regulatory region DNA binding
histone deacetylase activity
thiol-dependent ubiquitin-specific protease activity
protein binding
omega peptidase activity
ubiquitin protein ligase binding
identical protein binding
ATPase binding
Lys63-specific deubiquitinase activity
Lys48-specific deubiquitinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0