Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040000.2 | NP_001035089.1 | 11, 12 | 1715 | ||
NM_001207008.1 | NP_001193937.1 | 10, 11 | 1667 | |||
NM_001291964.1 | NP_001278893.1 | 11, 12 | 1769 | |||
XM_005266996.3 | XP_005267053.1 | 11, 12 | 1834 | |||
XM_005266997.2 | XP_005267054.1 | 11, 12 | 1832 | |||
XM_006715491.2 | XP_006715554.1 | 11, 12 | 1832 | |||
XM_006715494.3 | XP_006715557.1 | 10, 11 | 1769 | |||
XM_011535842.2 | XP_011534144.1 | 11, 12 | 1834 | |||
XM_017010878.1 | XP_016866367.1 | 11, 12 | 1832 | |||
XM_017010879.1 | XP_016866368.1 | 11, 12 | 1834 | |||
XM_017010880.1 | XP_016866369.1 | 11, 12 | 1813 | |||
XM_017010881.1 | XP_016866370.1 | 11, 12 | 1814 | |||
XM_017010882.1 | XP_016866371.1 | 11, 12 | 1832 | |||
XM_017010883.1 | XP_016866372.1 | 10, 11 | 1787 | |||
XM_017010884.1 | XP_016866373.1 | 10, 11 | 1789 | |||
XM_017010885.1 | XP_016866374.1 | 10, 11 | 1769 | |||
XM_017010886.1 | XP_016866375.1 | 11, 12 | 1832 | |||
XM_017010887.1 | XP_016866376.1 | 8, 9 | 1071 | |||
GenBank mRNA | AB209420.1 | - | 11, 12 | 1538 | ||
AB621809.1 | - | 11, 12 | 1769 | |||
AL161973.1 | - | 4, 5 | 571 | |||
BC014505.1 | - | 5, 6 | 668 | |||
U02478.1 | - | 10, 11 | 1525 |