Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002451.3 | NP_002442.2 | 4 | 311 | ||
GenBank mRNA | AF109294.1 | - | 4 | 319 | ||
AK300592.1 | - | 4 | 474 | |||
AK309365.1 | - | 4 | 298 | |||
AY712791.1 | - | 4 | 319 | |||
BC012316.1 | - | 4 | 285 | 3546198 | ||
BC018625.1 | - | 275 | 3030148 | |||
BC026106.1 | - | 4 | 293 | 4820938 | ||
CR541670.1 | - | 4 | 198 | |||
CR541710.1 | - | 4 | 198 | |||
FJ224338.1 | - | 4 | 198 | |||
HE654772.1 | - | 4 | 307 | |||
HE654773.1 | - | 4 | 307 | |||
HE654774.1 | - | 4 | 307 | |||
HE654775.1 | - | 4 | 307 | |||
HE654776.1 | - | 4 | 307 | |||
HE654777.1 | - | 4 | 307 | |||
L40432.1 | - | 4 | 308 | |||
U22233.1 | - | 4 | 319 |