Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTNR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005959.3 | NP_005950.1 | 1, 2 | 319 | ||
XM_011542839.2 | XP_011541141.1 | 1, 2 | 216 | |||
GenBank mRNA | AY521019.1 | - | 1, 2 | 216 | ||
BC069163.1 | - | 1, 2 | 319 | 7216982 | ||
U25341.1 | - | 1, 2 | 228 |
Set Membership: |
G-protein coupled receptor |