Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYBPC1 SIRNA Assays ›
Gene Symbol:
MYBPC1
Gene Name:
myosin binding protein C, slow type
Gene Aliases:
LCCS4, MYBPCC, MYBPCS
Chromosome Location:
Chr.12: 101594849 - 101695841 on Build GRCh38
UniGene ID:
Hs.654589
Species:
Human
siRNA ID
HSS106823
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MYBPC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001254718.1
NP_001241647.1
6
543
NM_001254719.1
NP_001241648.1
6
543
NM_001254720.1
NP_001241649.1
5
507
NM_001254721.1
NP_001241650.1
6
543
NM_001254722.1
NP_001241651.1
4
465
NM_001254723.1
NP_001241652.1
5
504
NM_002465.3
NP_002456.2
8
618
NM_206819.2
NP_996555.1
8
618
NM_206820.2
NP_996556.1
6
543
NM_206821.2
NP_996557.1
6
543
XM_005268876.4
XP_005268933.1
7
552
XM_006719405.3
XP_006719468.1
8
479
XM_006719406.3
XP_006719469.1
8
479
XM_006719407.3
XP_006719470.1
7
552
XM_006719408.3
XP_006719471.1
8
479
XM_006719409.3
XP_006719472.1
6
505
XM_006719410.3
XP_006719473.1
8
479
XM_006719411.3
XP_006719474.1
6
505
XM_017019315.1
XP_016874804.1
8
479
XM_017019316.1
XP_016874805.1
8
479
XM_017019317.1
XP_016874806.1
6
505
XM_017019318.1
XP_016874807.1
8
479
XM_017019319.1
XP_016874808.1
6
505
XM_017019320.1
XP_016874809.1
4
547
XM_017019321.1
XP_016874810.1
6
505
XM_017019322.1
XP_016874811.1
4
547
GenBank mRNA
AK056587.1
-
7
542
AK292976.1
-
8
578
AK295727.1
-
4
425
AK296681.1
-
6
507
AK303401.1
-
5
464
AL831993.2
-
8
553
AL832000.2
-
8
553
AL832332.2
-
8
553
AL832350.2
-
8
553
AL832354.2
-
8
555
AL832456.2
-
8
553
AL834249.2
-
8
553
BC015187.1
-
8
578
BC065720.1
-
4
428
BC092418.1
-
6
490
30529254
BC117217.1
-
4
400
40125768
BC143495.1
-
6
478
9052003
BC143502.1
-
4
400
9052010
BC143503.1
-
6
478
9052011
BC143504.1
-
5
442
9052012
BX499532.1
-
8
581
BX649073.2
-
9
649
CD614901.1
-
3
243
DA433791.1
-
6
507
X66276.1
-
6
496
X73114.1
-
6
484
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell adhesion
muscle filament sliding
positive regulation of protein targeting to mitochondrion
actin binding
protein binding
structural constituent of muscle
titin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7q954:80/10.239.162.42:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0