Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100112.1 | NP_001093582.1 | 29 | 4047 | ||
NM_017534.5 | NP_060004.3 | 29 | 4047 | |||
GenBank mRNA | AF111784.1 | - | 27 | 3919 | ||
BC126409.1 | - | 28 | 3990 | 8992125 | ||
BX510904.2 | - | 29 | 4010 |