Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO9A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006901.3 | NP_008832.2 | 25 | 4856 | ||
XM_006720539.2 | XP_006720602.1 | 25 | 4856 | |||
XM_011521613.2 | XP_011519915.1 | 25 | 4856 | |||
XM_011521614.2 | XP_011519916.1 | 26 | 4542 | |||
XM_011521615.2 | XP_011519917.1 | 25 | 4709 | |||
XM_011521616.2 | XP_011519918.1 | 25 | 4503 | |||
XM_011521617.2 | XP_011519919.1 | 25 | 4856 | |||
XM_011521618.2 | XP_011519920.1 | 25 | 4856 | |||
XM_011521619.2 | XP_011519921.1 | 24 | 4799 | |||
XM_011521620.2 | XP_011519922.1 | 24 | 4799 | |||
XM_011521621.2 | XP_011519923.1 | 24 | 4736 | |||
XM_011521622.2 | XP_011519924.1 | 25 | 4856 | |||
XM_011521623.2 | XP_011519925.1 | 8 | 1959 | |||
XM_017022228.1 | XP_016877717.1 | 25 | 4485 | |||
XM_017022229.1 | XP_016877718.1 | 26 | 4691 | |||
XM_017022230.1 | XP_016877719.1 | 24 | 4799 | |||
GenBank mRNA | AB209841.1 | - | 23 | 4235 | ||
AB290183.1 | - | 24 | 4291 | |||
AF117888.1 | - | 25 | 4607 | |||
AL137287.1 | - | 1 | 701 | |||
BC140869.1 | - | 25 | 4525 | 9021737 | ||
BC150294.1 | - | 23 | 3864 |