Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297418.1 | NP_001284347.1 | 6 | 821 | ||
NM_005968.4 | NP_005959.2 | 7 | 783 | |||
NM_031203.3 | NP_112480.2 | 8 | 666 | |||
XM_005272478.2 | XP_005272535.1 | 7 | 1127 | |||
XM_005272479.2 | XP_005272536.1 | 7 | 1127 | |||
XM_005272480.2 | XP_005272537.1 | 8 | 1010 | |||
XM_005272481.1 | XP_005272538.1 | 6 | 729 | |||
XM_005272483.2 | XP_005272540.1 | 7 | 594 | |||
XM_017026823.1 | XP_016882312.1 | 7 | 1127 | |||
XM_017026824.1 | XP_016882313.1 | 8 | 1010 | |||
XM_017026825.1 | XP_016882314.1 | 8 | 1010 | |||
XM_017026826.1 | XP_016882315.1 | 6 | 674 | |||
XM_017026827.1 | XP_016882316.1 | 6 | 729 | |||
XM_017026828.1 | XP_016882317.1 | 7 | 561 | |||
XM_017026829.1 | XP_016882318.1 | 7 | 612 | |||
XM_017026830.1 | XP_016882319.1 | 7 | 612 | |||
XM_017026831.1 | XP_016882320.1 | 7 | 322 | |||
XM_017026832.1 | XP_016882321.1 | 7 | 322 | |||
GenBank mRNA | AB209733.1 | - | 6 | 649 | ||
AF061832.1 | - | 8 | 816 | |||
AK024911.1 | - | 1 | 3524 | |||
AK309659.1 | - | 732 | ||||
BC000138.2 | - | 7 | 744 | 2900532 | ||
BC019580.1 | - | 7 | 712 | 4938259 | ||
L03532.1 | - | 7 | 711 | |||
L32611.1 | - | 8 | 595 | |||
X72018.1 | - | 3 | 349 |