Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000433.3 | NP_000424.2 | 14 | 1585 | ||
NM_001127651.2 | NP_001121123.1 | 15 | 1422 | |||
NM_001190789.1 | NP_001177718.1 | 12 | 1342 | |||
NM_001190794.1 | NP_001177723.1 | 13 | 1450 | |||
XM_005245207.1 | XP_005245264.1 | 13 | 1406 | |||
XM_011509580.1 | XP_011507882.1 | 15 | 1423 | |||
XM_011509581.1 | XP_011507883.1 | 15 | 1410 | |||
GenBank mRNA | AB209647.1 | - | 12 | 4855 | ||
AF527950.1 | - | 14 | 1332 | |||
AK296672.1 | - | 12 | 1198 | |||
AK298713.1 | - | 13 | 1359 | |||
AK312666.1 | - | 14 | 1495 | |||
BC001606.1 | - | 14 | 1562 | 3542841 | ||
BT007439.1 | - | 14 | 1310 | |||
KU178216.1 | - | 14 | 1310 | |||
KU178217.1 | - | 3 | 275 | |||
M32011.1 | - | 15 | 1377 |