Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NEB SIRNA Assays ›
Gene Symbol:
NEB
Gene Name:
nebulin
Gene Aliases:
NEB177D, NEM2
Chromosome Location:
Chr.2: 151485334 - 151734487 on Build GRCh38
UniGene ID:
Hs.588655
Species:
Human
siRNA ID
HSS107007
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NEB
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001164507.1
NP_001157979.1
60
8562
NM_001164508.1
NP_001157980.1
60
8562
NM_001271208.1
NP_001258137.1
60
8562
NM_004543.4
NP_004534.2
60
8562
XM_005246590.2
XP_005246647.1
60
8549
XM_005246591.2
XP_005246648.1
60
8549
XM_005246592.2
XP_005246649.1
60
8549
XM_005246593.2
XP_005246650.1
60
8549
XM_005246594.2
XP_005246651.1
60
8549
XM_005246596.2
XP_005246653.1
60
8549
XM_005246597.2
XP_005246654.1
60
8549
XM_005246598.2
XP_005246655.1
60
8549
XM_005246599.2
XP_005246656.1
60
8549
XM_005246600.2
XP_005246657.1
60
8549
XM_005246601.2
XP_005246658.1
60
8549
XM_005246602.2
XP_005246659.1
60
8549
XM_005246603.2
XP_005246660.1
60
8549
XM_005246604.2
XP_005246661.1
60
8549
XM_005246606.2
XP_005246663.1
60
8549
XM_005246608.2
XP_005246665.1
60
8549
XM_005246610.2
XP_005246667.1
60
8549
XM_005246611.2
XP_005246668.1
60
8549
XM_005246615.2
XP_005246672.1
60
8549
XM_005246616.1
XP_005246673.1
60
8549
XM_005246617.2
XP_005246674.1
60
8549
XM_006712541.2
XP_006712604.1
60
8549
XM_006712542.2
XP_006712605.1
60
8549
XM_011511225.2
XP_011509527.1
60
8549
XM_017004177.1
XP_016859666.1
60
8549
XM_017004178.1
XP_016859667.1
60
8549
XM_017004179.1
XP_016859668.1
60
8549
XM_017004180.1
XP_016859669.1
60
8549
XM_017004181.1
XP_016859670.1
60
8549
XM_017004182.1
XP_016859671.1
60
8549
XM_017004183.1
XP_016859672.1
60
8549
XM_017004184.1
XP_016859673.1
60
8549
XM_017004185.1
XP_016859674.1
60
8549
GenBank mRNA
X83957.1
-
47
8799
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
muscle organ development
somatic muscle development
muscle filament sliding
regulation of actin filament length
actin binding
protein binding
structural constituent of muscle
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0