Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKBIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020529.2 | NP_065390.1 | 1 | 236 | ||
GenBank mRNA | AK313421.1 | - | 1 | 232 | ||
AY033600.1 | - | 1 | 140 | |||
BC002601.2 | - | 1 | 228 | 3161686 | ||
BC004983.1 | - | 1 | 219 | 2957970 | ||
BT007091.1 | - | 1 | 126 | |||
M69043.1 | - | 1 | 220 |