Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005385.3 | NP_005376.2 | 16, 17 | 4449 | ||
XM_005265173.1 | XP_005265230.1 | 16, 17 | 4447 | |||
XM_006713171.1 | XP_006713234.1 | 17, 18 | 4366 | |||
XM_006713173.2 | XP_006713236.1 | 17, 18 | 4469 | |||
XM_011533747.2 | XP_011532049.1 | 17, 18 | 7781 | |||
XM_011533750.2 | XP_011532052.1 | 18, 19 | 4463 | |||
XM_017006474.1 | XP_016861963.1 | 17, 18 | 4363 | |||
XM_017006475.1 | XP_016861964.1 | 17, 18 | 4451 | |||
XM_017006476.1 | XP_016861965.1 | 11, 12 | 8994 | |||
XM_017006477.1 | XP_016861966.1 | 11, 12 | 5648 | |||
XM_017006478.1 | XP_016861967.1 | 11, 12 | 5625 | |||
XM_017006479.1 | XP_016861968.1 | 10, 11 | 6381 | |||
XM_017006480.1 | XP_016861969.1 | 18, 19 | 4456 | |||
GenBank mRNA | BC144518.1 | - | 12, 13 | 2904 | ||
BX957215.1 | - | 8, 9 | 5625 | |||
BX957218.1 | - | 18, 19 | 4477 | |||
CR749415.1 | - | 8, 9 | 5625 | |||
CR749571.1 | - | 18, 19 | 4477 | |||
L04288.2 | - | 16, 17 | 4490 |